Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 99.3% | 352.37 | 1 KG
Supplier: Medchemexpress LLC HYW0137721KG
Tris(4-methoxyphenyl)phosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is employed in Suzuki reaction and also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
- Biochemical reagent for life science research.
- Functions as a biological material or organic compound.
- Employed in Suzuki reaction.
- Used in kinetic studies of catalyst effects on biphenyl epoxy resin curing.