Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 99.3% | 352.37 | 25 G
SDP
Catalog No. 5000453238
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science related research. It is for research use only.
  • Used as a biological material or organic compound
  • Employed in Suzuki reaction
  • Used in the kinetic study of catalyst effects on biphenyl epoxy resin curing
  • For research use only
Catalog No. 50-004-53238 Supplier Medchemexpress LLC Supplier No. 25GHYW013772
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science related research. It is for research use only.
  • Used as a biological material or organic compound
  • Employed in Suzuki reaction
  • Used in the kinetic study of catalyst effects on biphenyl epoxy resin curing
  • For research use only