Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 99.3% | 352.37 | 50 G
SDP
Catalog No. 5000454198
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is employed in the Suzuki reaction and in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science-related research
Catalog No. 50-004-54198 Supplier Medchemexpress LLC Supplier No. HYW01377250G
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is employed in the Suzuki reaction and in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science-related research