Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | MFCD00014896 | 99.3% | 352.37 | 5 G
SDP
Catalog No. 5000454215
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been employed in the Suzuki reaction and used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Solid appearance
  • White to yellow color
  • Soluble in DMSO
  • Can be stored at 4°C under nitrogen
Catalog No. 50-004-54215 Supplier Medchemexpress LLC Supplier No. HYW0137725G
Tris(4-methoxyphenyl)phosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been employed in the Suzuki reaction and used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Solid appearance
  • White to yellow color
  • Soluble in DMSO
  • Can be stored at 4°C under nitrogen