Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III), 99%, Thermo Scientific™

Catalog No. AAA1416603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1416603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1416603 Supplier Thermo Scientific Chemicals Supplier No. A1416603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lewis catalyst for aldol reactions. NMR shift reagent.

Solubility
Soluble in hexane, methanol, tetrahydrofuran.

Notes
NMR Shift Reagent. Hygroscopic. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17978-77-7
Molecular Formula C30H33F21O6Pr
Molecular Weight (g/mol) 1029.462
MDL Number MFCD00064656
InChI Key LICWBBABBAOMPH-VNGPFPIXSA-N
Synonym praseodymium 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate
PubChem CID 131668219
IUPAC Name (Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one;praseodymium
SMILES CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)O.[Pr]

Specifications

Melting Point 215°C to 219°C
Quantity 1 g
Formula Weight 1026.45
Percent Purity 99%
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.