Learn More
Bio Rad Laboratories TRIS ACETATE
Catalog No. NC9028530
Encompass
Criterion XT Tris-Acetate Gel,30 microliter ,18-well,3-8%,shelf life of 8 months,for use with XT Tricine running buffer for separation of large proteins,can be used with Tris-glycine buffer when native PAGE conditions are required
Catalog No. NC9028530 Supplier Bio Rad Laboratories Supplier No. 3450130
Criterion XT Tris-Acetate Gel,30 microliter ,18-well,3-8%,shelf life of 8 months,for use with XT Tricine running buffer for separation of large proteins,can be used with Tris-glycine buffer when native PAGE conditions are required