Learn More
American Research Products Inc TRIS-ACETATE BUFFER, 0.2M,
Catalog No. NC1884734
Encompass
TRIS-Acetate Buffer, 0.2M, pH 7.8-500ml. 0.2M Tris base dissolved in 18.2 megohms water, pH adjusted to 7.8 with glacial acetic acid and filtered through 0.22-micron filter.
Catalog No. NC1884734 Supplier American Research Products Inc Supplier No. BB2412500ML
TRIS-Acetate Buffer, 0.2M, pH 7.8-500ml. 0.2M Tris base dissolved in 18.2 megohms water, pH adjusted to 7.8 with glacial acetic acid and filtered through 0.22-micron filter.