Learn More
Boston Bioproducts Inc Tris-Acetate Buffer (0.2 M, pH 7.4) - 500ml
Catalog No. NC1732330
Encompass
The Tris-Acetate Buffer is a buffering agent composed of Tris-base and acetic acid. The combination of these ingredients yields a pH range of 6.0-8.0. This buffer is ideal for requiring a pH slightly above physiological conditions. Often used to support the electrophoresis of nucleic acids on agarose gel and protein electrophoresis employing SDS-PAGE and related blotting techniques.
Catalog No. NC1732330 Supplier Boston Bioproducts Inc Supplier No. BB2410500ML
Add to Cart
Add to Cart
The Tris-Acetate Buffer is a buffering agent composed of Tris-base and acetic acid. The combination of these ingredients yields a pH range of 6.0-8.0. This buffer is ideal for requiring a pH slightly above physiological conditions. Often used to support the electrophoresis of nucleic acids on agarose gel and protein electrophoresis employing SDS-PAGE and related blotting techniques.