Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc TAE Buffer (10X), 1000 mL
SDP

Catalog No. 50983248
Encompass_Preferred

TAE (Tris-Acetate EDTA Buffer) buffer provides faster electrophoretic migration of linear DNA and better resolution of supercoiled DNA. TAE Buffer may be more suitable than TBE, for electrophoresis if DNA is to be recovered from the agarose and purified using commercially available DNA purification kits.

Applications: TAE is often prepared in concentrated stock solutions of 10X or 50X in the laboratory. A 1X working solution is prepared prior to electrophoresis.

Storage Condition: 15 °C to 30 °C
Shipping condition: ambient temperature
Sterile Filtered
DNase RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health

Catalog No. 50-983-248 Supplier Quality Biological Inc Supplier No. 351009131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAE (Tris-Acetate EDTA Buffer) buffer provides faster electrophoretic migration of linear DNA and better resolution of supercoiled DNA. TAE Buffer may be more suitable than TBE, for electrophoresis if DNA is to be recovered from the agarose and purified using commercially available DNA purification kits.

Applications: TAE is often prepared in concentrated stock solutions of 10X or 50X in the laboratory. A 1X working solution is prepared prior to electrophoresis.

Storage Condition: 15 °C to 30 °C
Shipping condition: ambient temperature
Sterile Filtered
DNase RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.