Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE Buffer, Tris-Acetate-EDTA, 25X Powder, Electrophoresis, Fisher BioReagents

Catalog No. BP13311 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1331-1 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP1331-1

missing translation for 'validMainframeMsg'

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 25X TAE Powder -- simply add pure water!
  • Each foil pack contains the correct amount of pre-weighed powder to make 1L of 25X TAE buffer containing 1M tris-acetate and 0.025M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 25X Powder
CAS 77-86-1, 6850-28-8, 6381-92-6, 10378-23-1
Color White
pH 8.3
Physical Form Solid
Quantity 1 Pk.
DNase DNase free
Packaging Foil Pack
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (64.36%), Acetic Acid, Sodium Salt (31.91%) and Ethylenediamine Tetraacetic Acid (3.72%).
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.