Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE Buffer, Tris-Acetate-EDTA, 25X Solution, Electrophoresis, Fisher BioReagents

Catalog No. BP13301 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1330-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP1330-1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-00-4

Nalgene Poly Bottle; 1L

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 25X TAE buffer solution - simply dilute as needed
  • 25X solution contains 1M tris-acetate and 0.025M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 24 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 25X Solution
CAS 60-00-4, 6850-28-8, 77-86-1, 7732-18-5
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (11.31%), EDTA Disodium Salt (0.87%), Acetic Acid, Sodium Salt (7.66%), and Hydrogen Chloride (3.74%).
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.