Learn More
Bioworld Tris-Acetate-SDS Running Buffer (10X), 500 mL
Catalog No. 50198648
Encompass_Preferred
TRIS-Acetate-SDS Running Buffer (10X) All bioWORLD running buffers are prepared with high quality reagents. Technical Information:Dilute with deionized water to make 1X. 10X concentration1X contains: 50mM Tris base, 50mM Tricine, 3.5mM SDS pH 8.24
Catalog No. 50-198-648 Supplier Bioworld Supplier No. 214200211
TRIS-Acetate-SDS Running Buffer (10X) All bioWORLD running buffers are prepared with high quality reagents. Technical Information:Dilute with deionized water to make 1X. 10X concentration1X contains: 50mM Tris base, 50mM Tricine, 3.5mM SDS pH 8.24