Learn More
Alkali Scientific Tris Acetate Ultra Pure, 1 Kg
SDP
Catalog No. 5000661687
Encompass_Preferred
Tris Acetate Ultra Pure is a high-purity buffering agent used in a variety of biochemical applications. This reagent is ideal for high-precision applications such as electrophoresis, protein analysis, and other laboratory techniques requiring a stable pH. The ultra-pure grade ensures minimal interference with sensitive experiments, maintaining the integrity of reactions. This 1kg bottle is perfect for large-scale buffer preparations, providing consistent buffering power across a range of experiments. Tris acetate is commonly used in molecular biology, immunology, and protein chemistry, particularly for its ability to maintain a stable pH in high-purity systems.
Catalog No. 50-006-61687 Supplier Alkali Scientific Supplier No. SAC002CP
Tris Acetate Ultra Pure is a high-purity buffering agent used in a variety of biochemical applications. This reagent is ideal for high-precision applications such as electrophoresis, protein analysis, and other laboratory techniques requiring a stable pH. The ultra-pure grade ensures minimal interference with sensitive experiments, maintaining the integrity of reactions. This 1kg bottle is perfect for large-scale buffer preparations, providing consistent buffering power across a range of experiments. Tris acetate is commonly used in molecular biology, immunology, and protein chemistry, particularly for its ability to maintain a stable pH in high-purity systems.