Learn More
Alkali Scientific Tris Acetate Ultra Pure, 100 G
SDP
Catalog No. 5000661738
Encompass_Preferred
Tris Acetate Ultra Pure is a high-purity buffering agent used in laboratory applications where a stable pH is required. The 100g size is suitable for preparing buffer solutions in a variety of molecular biology and biochemistry experiments. Tris Acetate is particularly useful in gel electrophoresis for nucleic acids, where it provides an optimal buffering capacity to stabilize the pH during the separation process. The high purity of this reagent ensures that it does not interfere with the analysis of proteins or nucleic acids, making it ideal for high-precision experiments. This buffering agent is widely used in research and diagnostics where reliable pH control is essential for reproducible results.
Catalog No. 50-006-61738 Supplier Alkali Scientific Supplier No. SAC001CP
Tris Acetate Ultra Pure is a high-purity buffering agent used in laboratory applications where a stable pH is required. The 100g size is suitable for preparing buffer solutions in a variety of molecular biology and biochemistry experiments. Tris Acetate is particularly useful in gel electrophoresis for nucleic acids, where it provides an optimal buffering capacity to stabilize the pH during the separation process. The high purity of this reagent ensures that it does not interfere with the analysis of proteins or nucleic acids, making it ideal for high-precision experiments. This buffering agent is widely used in research and diagnostics where reliable pH control is essential for reproducible results.