Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Acetonitrile)Cyclopentadienylruthenium(II) Hexafluorophosphate

Catalog No. 673307250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-330-7250MG 250 mg
67-330-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 67-330-7250MG Supplier Sigma Aldrich Supplier No. 673307250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Regioselective and stereoselective dehydrative allylation of nucleophilesO-H Insertion and condensation reactionsHydrosilylationDecarboxylative allylic etherificationClaisen rearrangement of unactivated allyl vinyl ethers

Specifications

CAS 80049-61-2
Molecular Formula C11H14F6N3PRu
Linear Formula [(C5H5)Ru(CH3CN)3]PF6
MDL Number MFCD02684569
Quantity 250 mg
Synonym (Cyclopentadienyltris(acetonitrile)ruthenium hexafluorophosphate; Tris(acetonitrile)cyclopentadienylruthenium hexafluorophosphate
Molecular Weight (g/mol) 434.28 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.