Learn More
Strem, An Ascensus Company CAS# 59738-27-1. 1g. Tris(acetonitrile)pentamethylcyclopentadienylrhodium(III) hexafluoroantimonate, min. 98%. MFCD18827646
SDP
Supplier:  Strem, An Ascensus Company 4521601G
Encompass
CAS# 59738-27-1. 1g. Tris(acetonitrile)pentamethylcyclopentadienylrhodium(III) hexafluoroantimonate, min. 98%. MFCD18827646. Molecular Weight: 832.79. Molecular Formula: [Rh(C10H15)(CH3CN)3]+(SbF6ae)2. Color/form: light yellow pwdr. Strem# 45-2160. http://www.strem.com/catalog/v/45-2160/
Catalog No. 50-373-5