Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS ACS Ultrapure, Lab Chem

Catalog No. LC262701
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
10 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC262701 500 g
LC262703 2.5 kg
LC262702 1 kg
LC262706K 5 kg
LC262705K 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC262701 Supplier LabChem, Inc. Supplier No. LC262701
Add to Cart
Add to Cart
  • TRIS ACS Ultrapure Analytical Standard

Specifications

Chemical Name or Material Tris(hydroxymethl)aminomethane
Melting Point 175 °C
CAS 77-86-1
Boiling Point 219 °C
Color White
Decomposition Information Stable
Density 1.35 g/cm3
For Use With (Application) Laboratory
For Use With (Equipment) Instruments and Electrodes
pH 10.0 to 11.5 for 5% solution
Physical Form Crystalline Powder
Quantity 500 g
Solution Type Standard
Flash Point Does not flash
Assay Percent Range 99.8% to 100.2%
Infrared Spectrum Passes Test
Molecular Formula NH2C(CH2OH)3
Linear Formula NH2C(CH2OH)3
Packaging Plastic Bottle
Specific Gravity 1.35
Sterility Non-sterile
Merck Index 1,309,842
Solubility Information Soluble
IUPAC Name Tris(hydroxymethyl)aminomethane
Molecular Weight (g/mol) 121.135
Formula Weight 121.135
Percent Purity 99.8% to 100.2%
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.