Learn More
Alkali Scientific TRIS Amino Ultra PC [Tris (Hydroxymethyl) Aminomethane] [Tromethamine], 10 Kg
SDP
Catalog No. 5000659920
Encompass_Preferred
TRIS Amino Ultra PC [Tris (Hydroxymethyl) Aminomethane] is a high-purity reagent commonly used in biochemical research and laboratory applications. This 10kg package offers an ideal supply for large laboratories or commercial research facilities requiring a stable and reliable source of Tris Amino for buffer preparation. Tris Amino is used extensively in applications where precise pH control is necessary, such as in protein purification, nucleic acid analysis, and electrophoresis. Its ultra-pure quality ensures minimal contamination, making it suitable for sensitive applications in molecular biology, biochemistry, and other disciplines requiring high-quality reagents for accurate experimentation.
Catalog No. 50-006-59920 Supplier Alkali Scientific Supplier No. ISA001CP
TRIS Amino Ultra PC [Tris (Hydroxymethyl) Aminomethane] is a high-purity reagent commonly used in biochemical research and laboratory applications. This 10kg package offers an ideal supply for large laboratories or commercial research facilities requiring a stable and reliable source of Tris Amino for buffer preparation. Tris Amino is used extensively in applications where precise pH control is necessary, such as in protein purification, nucleic acid analysis, and electrophoresis. Its ultra-pure quality ensures minimal contamination, making it suitable for sensitive applications in molecular biology, biochemistry, and other disciplines requiring high-quality reagents for accurate experimentation.