Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma base, 77-86-1, MFCD00004679, 100 g
Catalog No. NC1246588
Encompass
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration), Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol.
Catalog No. NC1246588 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T6066100G
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration), Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol.