Learn More
Boston Bioproducts Inc Tris-Base (2 M, pH > 10.0) - 250ml
Catalog No. NC2182790
Encompass
Tris-base is a widely used biological buffer in various research applications in biochemistry, molecular biology, and cell biology. The buffering capabilities of Tris-base are effective in the pH range of 7.0-9.0, making it suited for various biological applications. This buffer is widely used in protocols for DNA and RNA extraction, in situ hybridization, protein extraction from cells, in agarose gel electrophoresis, polyacrylamide gel electrophoresis and as a wash buffer in Western blotting.
This item is not returnable. View return policy
Catalog No. NC2182790 Supplier Boston Bioproducts Inc Supplier No. BB2689250ML
Tris-base is a widely used biological buffer in various research applications in biochemistry, molecular biology, and cell biology. The buffering capabilities of Tris-base are effective in the pH range of 7.0-9.0, making it suited for various biological applications. This buffer is widely used in protocols for DNA and RNA extraction, in situ hybridization, protein extraction from cells, in agarose gel electrophoresis, polyacrylamide gel electrophoresis and as a wash buffer in Western blotting.