Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris Base, 80g, CAS No.: 77-86-1
Catalog No. NC3705411
Encompass
Synonym(s): 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol. Linear Formula: NH2C(CH2OH)3
Catalog No. NC3705411 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1024080083
Synonym(s): 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol. Linear Formula: NH2C(CH2OH)3