Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS (Base), BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog No. 02004510 Shop All Avantor Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-510 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-510 Supplier Avantor Supplier No. 409906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality biochemicals for research use

    For Liquid Chromatography Applications

Specifications

CAS 77-86-1
Packaging Poly Pail
Molecular Formula C4H11NO3
Quantity 5 kg
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Formula Weight 121.14
Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material TRIS (Base)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.