Learn More
Pfanstiehl Inc Tris Base (Tromethamine) High Purity, Low Endotoxin, Low Metals, USP, EP, BP, ChP (T-153) 100G GMP, Multi-compendial CAS: 77-86-1 Injectable Excipient Grade
Catalog No. NC2953466 Shop All Pfanstiehl Products
Encompass
Pfanstiehl Tris Base, High Purity, Low Endotoxin, Low Metals, USP, EP, BP, ChP is a multi-compendial buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Tris Base meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl Brand Tris Base is compliant with USP (United States Pharmacopoeia) EP (European Pharmacopoeia), BP (British Pharmacopeia) and ChP (Chinese Pharmacopeia) Compendial, U.S.P. , cGMP , pharmaceutical grade , multicompendial
Catalog No. NC2953466 Supplier Pfanstiehl Inc Supplier No. T153RD
Pfanstiehl Tris Base, High Purity, Low Endotoxin, Low Metals, USP, EP, BP, ChP is a multi-compendial buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Tris Base meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl Brand Tris Base is compliant with USP (United States Pharmacopoeia) EP (European Pharmacopoeia), BP (British Pharmacopeia) and ChP (Chinese Pharmacopeia) Compendial, U.S.P. , cGMP , pharmaceutical grade , multicompendial