Learn More
Alkali Scientific Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 25 Kg
SDP
Catalog No. 5000659970
Encompass_Preferred
Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 25kg, is a high-purity chemical reagent used in a variety of biochemical and molecular biology applications. This bulk-size package is ideal for laboratories or industries that require large volumes of Tris Base for buffer preparation, research, or manufacturing processes. Tris Base is used to prepare Tris-buffered saline (TBS) solutions, a common buffer used in molecular biology, electrophoresis, and enzyme analysis. With a neutral pH, it offers consistent pH control, which is essential for maintaining the stability of proteins and nucleic acids in solution. This large-size package ensures an uninterrupted supply for laboratories performing extensive research or commercial production.
Catalog No. 50-006-59970 Supplier Alkali Scientific Supplier No. ISB005CP
Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 25kg, is a high-purity chemical reagent used in a variety of biochemical and molecular biology applications. This bulk-size package is ideal for laboratories or industries that require large volumes of Tris Base for buffer preparation, research, or manufacturing processes. Tris Base is used to prepare Tris-buffered saline (TBS) solutions, a common buffer used in molecular biology, electrophoresis, and enzyme analysis. With a neutral pH, it offers consistent pH control, which is essential for maintaining the stability of proteins and nucleic acids in solution. This large-size package ensures an uninterrupted supply for laboratories performing extensive research or commercial production.