Learn More
Alkali Scientific Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 250 G
SDP
Supplier:  Alkali Scientific ISB004CP
Encompass_Preferred
Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 250g, is a high-quality reagent commonly used in biochemical, molecular biology, and analytical research. This ultra-pure form of Tris Base ensures low levels of impurities, making it ideal for sensitive applications such as electrophoresis, protein analysis, and enzyme reactions. The 250g size is suitable for smaller laboratories or for preparing smaller volumes of Tris-buffered solutions. Tris is essential for stabilizing the pH of reaction mixtures, making it widely used in buffer formulations and for preparing buffers in protein and nucleic acid research. It is an essential reagent for various applications requiring precise pH control.
Catalog No. 50-006-59972