Learn More
AdipoGen Tris-bisCF3phenylphosphine
SDP
Supplier:  AdipoGen CDXT0049M500
Encompass_Preferred
Chemical. CAS 175136-62-6. Formula C24H9F18P. MW 670.27. Synthetic A phosphine with enhanced solubility in solvents. Phosphines are used as ligands for many metal complexes.
Catalog No. 50-256-0029