Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Borate EDTA Buffer

Catalog No. BMA51234 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA51234 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA51234 Supplier Lonza Inc Supplier No. 51234
Add to Cart
Add to Cart

Chemical Identifiers

CAS 610769-35-2
Molecular Formula C14H30BN3O14
Molecular Weight (g/mol) 475.211
InChI Key OSBLTNPMIGYQGY-UHFFFAOYSA-N
PubChem CID 21257725
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;boric acid
SMILES B(O)(O)O.C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.C(C(CO)(CO)N)O

Specifications

Quantity 1 L
Chemical Name or Material Tris borate EDTA Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.