Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris-Borate-EDTA buffer Bi
Catalog No. 502900049
Encompass
TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This SOLUTIONon is effective under slightly basic conditions which keeps DNA deprotonated water-soluble and protected from degradation. This concentrate can be easily diluted to 1x or 0.5x before use (with molecular biology grade water).
Catalog No. 50-290-0049 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T44154L
TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This SOLUTIONon is effective under slightly basic conditions which keeps DNA deprotonated water-soluble and protected from degradation. This concentrate can be easily diluted to 1x or 0.5x before use (with molecular biology grade water).