Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-Borate-EDTA Buffer, MP Biomedicals™

Catalog No. MP04816721 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04816721 1 L
MP04816724 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04816721 Supplier MP Biomedicals, Inc Supplier No. 04816721
Add to Cart
Add to Cart

Tris-Borate-EDTA Buffer, Clear, Colorless Solution

  • TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis.
  • TBE is used with non-denaturing or denaturing (7 M urea) gels.
  • It is also routinely used for DNA automated sequencing gel.
  • TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
  • It is used to prepare a 1 X buffer for polyacrylamide and agarose gel electrophoresis.
  • TBE buffer is a solution of 1M Tris, 0.9M boric acid and 0.01M EDTA disodium salt, prepared from MP's Ultra Pure products.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00236358
Synonym TBE buffer

Specifications

pH 8.3 to 8.5 (as is); 8.23 to 8.43 (1X)
Physical Form Solution
Quantity 1 L
storageNote1 Store at Room Temperature(15-30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.