Learn More
Biotang Inc Tris-Buffered Saline 10X pH 7.6 (BM-615): 250 mM Tris, 27 mM KCl, 1.37 M NaCl, pH 7.6 +/- 0.2. Prepared in 18.2 megohms water. To make 1X, dilute 10-fold with deionized water. The pH of the 1X should be 7.6 +/- 0.2.
Catalog No. 50155734 Shop All BIOTANG Inc Products
Encompass_Preferred
Tris-Buffered Saline 10X pH 7.6 (BM-615): 250 mM Tris, 27 mM KCl, 1.37 M NaCl, pH 7.6 +/- 0.2. Prepared in 18.2 megohms water. To make 1X, dilute 10-fold with deionized water. The pH of the 1X should be 7.6 +/- 0.2.
Catalog No. 50-155-734 Supplier Biotang Inc Supplier No. BTBB184
Tris-Buffered Saline 10X pH 7.6 (BM-615): 250 mM Tris, 27 mM KCl, 1.37 M NaCl, pH 7.6 +/- 0.2. Prepared in 18.2 megohms water. To make 1X, dilute 10-fold with deionized water. The pH of the 1X should be 7.6 +/- 0.2.