Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphate Buffered Saline with 0.05% TWEEN 20, pH 7.4, 10 Pack
Catalog No. NC1500806
Encompass
Phosphate Buffered Saline with 0.05% TWEEN 20, pH 7.4, 10 Pack
Catalog No. NC1500806 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P356310PAK
Phosphate Buffered Saline with 0.05% TWEEN 20, pH 7.4, 10 Pack