Learn More
Kd Medical Inc TRIS BUFF SALINE,10X PH 7.6 1L
SDP
Catalog No. 501031416
Encompass_Preferred
Commonly used buffer used as a wash buffer in multiple laboratory applications. 10X concentrated TBS is ready to be diluted to a 1X working solution. pH 7.6, one 1 Liter bottle.
Catalog No. 50-103-1416 Supplier Kd Medical Inc Supplier No. RGF3345
Commonly used buffer used as a wash buffer in multiple laboratory applications. 10X concentrated TBS is ready to be diluted to a 1X working solution. pH 7.6, one 1 Liter bottle.