Learn More
Electron Microscopy Sciences Tris Buffer 0.2M, pH 8.0 250 ML
SDP
Catalog No. 501901211
Encompass_Preferred
C₄H₁₁NO₃ F.W. 121.14
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
pH: 8.0
Catalog No. 50-190-1211 Supplier Electron Microscopy Sciences Supplier No. 1173005
C₄H₁₁NO₃ F.W. 121.14
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
pH: 8.0