Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Buffer, 0.5 L, MilliporeSigma™

Catalog No. LSKCTB500MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKCTB500MI 0.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKCTB500MI Supplier MilliporeSigma Supplier No. LSKCTB500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable storage buffer for plasmid and BAC minipreps.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
For Use With (Application) Laboratory
Physical Form Liquid
Quantity 0.5 L
Solution Type Buffer, Storage
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.