Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tris Buffer, 100mM, pH 7.4, Molecular Biology Grade, Calbiochem™,

Catalog No. 648315100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-831-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-831-5100ML Supplier MilliporeSigma™ Supplier No. 648315100ML

missing translation for 'validMainframeMsg'

Useful in the pH range of 10.2 to 10.6

Widely used biological buffer, Suitable for DNA and RNA applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym tris(Hydroxymethyl)aminomethane, HCl
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Boiling Point 219°C to 220°C
Quantity 100 mL
DNase None Detected
Solubility Information Soluble in water (800g/L at 20°C)
Formula Weight 121.1
Grade Molecular Biology
Chemical Name or Material Tris Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.