Learn More
Kd Medical Inc TRIS BUFFER, 2M,PH 8.0, BTL
SDP
Catalog No. 501031292
Encompass_Preferred
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers> One 500 ml bottle.
Catalog No. 50-103-1292 Supplier Kd Medical Inc Supplier No. RGE3380
Add to Cart
Add to Cart
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers> One 500 ml bottle.