Learn More
Crescent Chemical Co Inc TRIS BUFFER PH 8.8 1 M SOLN
SDP
Catalog No. 50248481
Encompass_Preferred
Tris buffer pH 8.8, 1 M solution molecular biology grade DNase, RNase, Protease not detected. The pH value of Tris buffer is temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03 - 0.05 unit
Catalog No. 50-248-481 Supplier Crescent Chemical Co Inc Supplier No. SE39794.01
Add to Cart
Add to Cart
Tris buffer pH 8.8, 1 M solution molecular biology grade DNase, RNase, Protease not detected. The pH value of Tris buffer is temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03 - 0.05 unit