Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris Buffered Saline, MFCD00132476, 10pk
Catalog No. NC0763525
Encompass
pH 8.0, powder, Synonym: TBS. Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.05 M Tris buffered saline (NaCl - 0.138 M; KCl - 0.0027 M); pH 8.0, at 25 °C.
Catalog No. NC0763525 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T666410PAK
pH 8.0, powder, Synonym: TBS. Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.05 M Tris buffered saline (NaCl - 0.138 M; KCl - 0.0027 M); pH 8.0, at 25 °C.