Learn More
Lonza Walkersville Tris Buffer 50 mM
Catalog No. NC9496398
Encompass
Tris buffer can be used in place of water as the sample diluent for highly acidic or basic samples (for endotoxin testing, samples and sample dilutions should be between pH 6-8). Tris buffer may be used to prepare samples for any of our endotoxin detection assays. Contains <0.005 EU/ml endotoxin.Storage: 2°C - 8°CContent:1 vial
Catalog No. NC9496398 Supplier Lonza Walkersville Supplier No. S50642
Tris buffer can be used in place of water as the sample diluent for highly acidic or basic samples (for endotoxin testing, samples and sample dilutions should be between pH 6-8). Tris buffer may be used to prepare samples for any of our endotoxin detection assays. Contains <0.005 EU/ml endotoxin.Storage: 2°C - 8°CContent:1 vial