Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris Buffered Saline, MFCD00132476, 1 L
Catalog No. NC1503432
Encompass
Synonym: TBS, 10 ×, solution. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash microtiter plates for western blot experiments and screening methods.
Catalog No. NC1503432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T59121L
Synonym: TBS, 10 ×, solution. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash microtiter plates for western blot experiments and screening methods.