Learn More
Medchemexpress LLC Tris buffered saline | 10 TABLETS
SDP
Catalog No. 5000284356
Encompass_Preferred
Tris buffered saline is supplied as ready-to-dissolve tablets for preparing Tris/NaCl buffer solutions commonly used in molecular biology and protein applications. The tablet format simplifies buffer preparation and storage while delivering a stable pH environment for assays and electrophoresis.
  • Ready-to-use tablet format for convenient buffer preparation.
  • One tablet dissolves in 15 mL deionized water to yield 0.05 M Tris and 0.15 M sodium chloride.
  • pH approximately 7.6 at 25 °C.
  • White to off-white solid tablets for easy visual quality check.
  • Suitable for DNA/RNA extraction, PCR, and protein electrophoresis.
  • Compact packaging reduces storage space and eases transport.
Catalog No. 50-002-84356 Supplier Medchemexpress LLC Supplier No. HYW46047110TABLETS
Tris buffered saline is supplied as ready-to-dissolve tablets for preparing Tris/NaCl buffer solutions commonly used in molecular biology and protein applications. The tablet format simplifies buffer preparation and storage while delivering a stable pH environment for assays and electrophoresis.
  • Ready-to-use tablet format for convenient buffer preparation.
  • One tablet dissolves in 15 mL deionized water to yield 0.05 M Tris and 0.15 M sodium chloride.
  • pH approximately 7.6 at 25 °C.
  • White to off-white solid tablets for easy visual quality check.
  • Suitable for DNA/RNA extraction, PCR, and protein electrophoresis.
  • Compact packaging reduces storage space and eases transport.