Learn More
Bioworld Tris Buffered Saline, 500 mL
Catalog No. 50255485
Encompass_Preferred
Tris Buffered SalineContains: 0.1M Tris and 0.3M NaCl, pH 7.5
Catalog No. 50-255-485 Supplier Bioworld Supplier No. 402200941
Tris Buffered SalineContains: 0.1M Tris and 0.3M NaCl, pH 7.5