Learn More
Boston Bioproducts Inc Tris Buffered Saline, TBS (10X, pH 7.4, Low Endotoxin) - 100ml
Catalog No. NC2089953
Encompass
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical immunoassay as a wash buffer. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 10X concentration and tested for endotoxin.
This item is not returnable. View return policy
Catalog No. NC2089953 Supplier Boston Bioproducts Inc Supplier No. BM300LT100ML
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical immunoassay as a wash buffer. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 10X concentration and tested for endotoxin.