Learn More
Alkali Scientific TRIS Buffered Saline, 10X Solution, 1 L
SDP
Catalog No. 5000661140
Encompass_Preferred
TRIS Buffered Saline (TBS) 10X solution is a concentrated buffer used in various biological applications, including immunohistochemistry, protein assays, and cell biology research. This 1L bottle of 10X solution is designed for dilution to 1X working concentration, making it convenient for a wide range of protocols. TBS provides a stable, isotonic environment that is ideal for washing cells, diluting samples, and preparing buffers for antibody conjugation. The buffering capacity of TRIS ensures stable pH conditions, making TBS essential for maintaining protein and enzyme functionality during experiments. It is commonly used for western blotting, enzyme-linked immunosorbent assays (ELISA), and other immunological procedures.
Catalog No. 50-006-61140 Supplier Alkali Scientific Supplier No. TRB002CP
Add to Cart
Add to Cart
TRIS Buffered Saline (TBS) 10X solution is a concentrated buffer used in various biological applications, including immunohistochemistry, protein assays, and cell biology research. This 1L bottle of 10X solution is designed for dilution to 1X working concentration, making it convenient for a wide range of protocols. TBS provides a stable, isotonic environment that is ideal for washing cells, diluting samples, and preparing buffers for antibody conjugation. The buffering capacity of TRIS ensures stable pH conditions, making TBS essential for maintaining protein and enzyme functionality during experiments. It is commonly used for western blotting, enzyme-linked immunosorbent assays (ELISA), and other immunological procedures.