Learn More
Alkali Scientific TRIS Buffered Saline, 10X Solution, 4 L
SDP
Catalog No. 5000661500
Encompass_Preferred
TRIS Buffered Saline (TBS) 10X solution is a concentrated buffer commonly used in molecular biology, immunology, and biochemistry. This 4L bottle provides a convenient, large-volume supply of buffer for high-throughput laboratories and research facilities. TBS is widely used in protocols such as washing cells, diluting proteins, and preparing buffers for antibody conjugation in applications like western blotting and ELISA. The TRIS component ensures that the pH remains stable, preventing fluctuations that could affect the accuracy of experimental results. This 10X solution is easily diluted to 1X, making it ideal for a variety of laboratory applications where stable pH and ionic strength are essential.
Catalog No. 50-006-61500 Supplier Alkali Scientific Supplier No. TRB004CP
Add to Cart
Add to Cart
TRIS Buffered Saline (TBS) 10X solution is a concentrated buffer commonly used in molecular biology, immunology, and biochemistry. This 4L bottle provides a convenient, large-volume supply of buffer for high-throughput laboratories and research facilities. TBS is widely used in protocols such as washing cells, diluting proteins, and preparing buffers for antibody conjugation in applications like western blotting and ELISA. The TRIS component ensures that the pH remains stable, preventing fluctuations that could affect the accuracy of experimental results. This 10X solution is easily diluted to 1X, making it ideal for a variety of laboratory applications where stable pH and ionic strength are essential.