Learn More
Boston Bioproducts Inc Tris Buffered Saline, TBS (20X, for Western Blot Washing) - 1L
Catalog No. NC2066510
Encompass
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.
Catalog No. NC2066510 Supplier Boston Bioproducts Inc Supplier No. BM301X1L
Add to Cart
Add to Cart
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.