Learn More
Boston Bioproducts Inc Tris Buffered Saline, TBS (20X) - 4L
Catalog No. NC1411925
Encompass
The Tris Buffered Saline (TBS) is an isotonic solution frequently used in biological research due to its non-toxic nature. The formulation of TBS includes Tris (tris(hydroxymethyl)aminomethane), which serves as a buffering agent, and Sodium Chloride (NaCl) salt. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.
Catalog No. NC1411925 Supplier Boston Bioproducts Inc Supplier No. BM300X4L
The Tris Buffered Saline (TBS) is an isotonic solution frequently used in biological research due to its non-toxic nature. The formulation of TBS includes Tris (tris(hydroxymethyl)aminomethane), which serves as a buffering agent, and Sodium Chloride (NaCl) salt. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.