Learn More
Research Products International Corp TRIS, Buffered Saline, 500 ML
SDP
Catalog No. 502230480
Encompass_Preferred
TBS 10X Solution (Tris Buffered Saline) is a concentrated buffered salt solution containing Tris and sodium chloride. It maintains pH stability and provides ionic strength for biological experiments. Tris acts as a buffering agent to keep pH consistent, while sodium chloride mimics physiological conditions. Dilute 10X solution with distilled water to prepare working concentrations (e.g., 1X). TBS is widely used in protein and nucleic acid blotting, immunohistochemistry, and molecular biology techniques, ensuring stable and reproducible results across diverse applications.
Catalog No. 50-223-0480 Supplier Research Products International Corp Supplier No. T60075500.0
TBS 10X Solution (Tris Buffered Saline) is a concentrated buffered salt solution containing Tris and sodium chloride. It maintains pH stability and provides ionic strength for biological experiments. Tris acts as a buffering agent to keep pH consistent, while sodium chloride mimics physiological conditions. Dilute 10X solution with distilled water to prepare working concentrations (e.g., 1X). TBS is widely used in protein and nucleic acid blotting, immunohistochemistry, and molecular biology techniques, ensuring stable and reproducible results across diverse applications.