Learn More
Bioworld Tris Buffered Saline, 1 L
Catalog No. 50255486
Encompass_Preferred
Tris Buffered SalineContains: 0.1M Tris and 0.3M NaCl, pH 7.5
Catalog No. 50-255-486 Supplier Bioworld Supplier No. 402200942
Tris Buffered SalineContains: 0.1M Tris and 0.3M NaCl, pH 7.5