Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc TBS (10X), pH 7.4, 500 mL
SDP

Catalog No. 50983267
Encompass_Preferred

TBS (10X), pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.

DNase, RNase and Protease tested.

Applications: TBS (10X), pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS (10X), pH 7.44 can also be used in detergent preparation and Western blot or ELISA applications.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
pH: 7.4 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-267 Supplier Quality Biological Inc Supplier No. 351086101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBS (10X), pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.

DNase, RNase and Protease tested.

Applications: TBS (10X), pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS (10X), pH 7.44 can also be used in detergent preparation and Western blot or ELISA applications.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
pH: 7.4 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.